Assay Details
| Gene Symbol: igsf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110473.1 | NP_001103943.1 | 3 - 4 | 482 | 65 | |
| GenBank mRNA | BC134916.1 | - | 3 - 4 | 367 | 65 | |
| CU458945.1 | - | 3 - 4 | 512 | 65 | ||
| KM655674.1 | - | 3 - 4 | 450 | 65 | ||
More Information
Set Membership: |
|

igsf21b
Genomic Map