Assay Details
| Gene Symbol: igsf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110473.1 | NP_001103943.1 | 7 - 8 | 1304 | 72 | |
| XM_017353589.2 | XP_017209078.1 | 6 - 7 | 1050 | 72 | ||
| GenBank mRNA | BC134916.1 | - | 7 - 8 | 1189 | 72 | |
| CU458945.1 | - | 7 - 8 | 1334 | 72 | ||
| KM655674.1 | - | 7 - 8 | 1272 | 72 | ||
More Information
Set Membership: |
|

igsf21b
Genomic Map