Assay Details
| Gene Symbol: igsf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110473.1 | NP_001103943.1 | 8 - 9 | 1492 | 91 | |
| XM_017353589.2 | XP_017209078.1 | 7 - 8 | 1238 | 91 | ||
| GenBank mRNA | BC134916.1 | - | 8 - 9 | 1377 | 91 | |
| CU458945.1 | - | 8 - 9 | 1522 | 91 | ||
| KM655674.1 | - | 8 - 9 | 1460 | 91 | ||
More Information
Set Membership: |
|

igsf21b
Genomic Map