Assay Details
| Gene Symbol: ghrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111081.2 | NP_001104551.1 | 4 - 5 | 764 | 116 | |
| GenBank mRNA | EU649775.1 | - | 4 - 5 | 116 | ||
More Information
Set Membership: |
|

ghrb
Genomic Map