Assay Details
Gene Symbol: ghrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111081.2 | NP_001104551.1 | 6 - 7 | 1100 | 86 | |
GenBank mRNA | EU649775.1 | - | 6 - 7 | 1103 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
