Assay Details
| Gene Symbol: ghrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111081.2 | NP_001104551.1 | 8 - 9 | 1259 | 93 | |
| GenBank mRNA | GQ890153.1 | - | 2 - 3 | 149 | 93 | |
| GQ890154.1 | - | 2 - 3 | 149 | 93 | ||
More Information
Set Membership: |
|

ghrb
Genomic Map