Assay Details
| Gene Symbol: hpxa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328534.1 | NP_001315463.1 | 5 - 6 | 732 | 80 | |
| GenBank mRNA | BC056563.1 | - | 5 - 6 | 561 | 80 | |
| BC155107.1 | - | 7 - 8 | 944 | 80 | ||
More Information
Set Membership: |
|

hpxa
Genomic Map