Assay Details
| Gene Symbol: hpxa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328534.1 | NP_001315463.1 | 8 - 9 | 1162 | 73 | |
| GenBank mRNA | BC056563.1 | - | 8 - 9 | 991 | 73 | |
| BC155107.1 | - | 10 - 11 | 1374 | 73 | ||
More Information
Set Membership: |
|

hpxa
Genomic Map