Assay Details
| Gene Symbol: etaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111157.1 | NP_001104627.1 | 1 - 2 | 223 | 105 | |
| XM_009294369.2 | XP_009292644.1 | 2 - 3 | 521 | 105 | ||
| GenBank mRNA | BC153950.1 | - | 1 - 2 | 223 | 105 | |
More Information
Set Membership: |
|

etaa1
Genomic Map