Assay Details
Gene Symbol: dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111169.1 | NP_001104639.1 | 3 - 4 | 307 | 77 | |
XM_005160743.4 | XP_005160800.1 | 3 - 4 | 416 | 77 | ||
XM_005160744.4 | XP_005160801.1 | 3 - 4 | 416 | 77 | ||
XM_009294764.3 | XP_009293039.1 | 3 - 4 | 416 | 77 | ||
GenBank mRNA | EU245032.1 | - | 3 - 4 | 307 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
