Assay Details
| Gene Symbol: etf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111171.1 | NP_001104641.1 | 109 | 109 | ||
| GenBank mRNA | BC153983.1 | - | 109 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC153983.1, NM_001111171.1 transcripts.
Set Membership: |
|

etf1a
Genomic Map