Assay Details
| Gene Symbol: htra1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111182.1 | NP_001104652.1 | 1 - 2 | 588 | 71 | |
| GenBank mRNA | BC155591.1 | - | 1 - 2 | 588 | 71 | |
More Information
Set Membership: |
|

htra1b
Genomic Map