Assay Details
| Gene Symbol: prph2lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111194.1 | NP_001104664.1 | 2 - 3 | 911 | 78 | |
| XM_005160581.4 | XP_005160638.1 | 3 - 4 | 1301 | 78 | ||
| GenBank mRNA | BC155639.1 | - | 2 - 3 | 911 | 78 | |
More Information
Set Membership: |
|

prph2lb
Genomic Map