Assay Details
| Gene Symbol: shrprbck1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111200.1 | NP_001104670.1 | 11 - 12 | 2030 | 94 | |
| GenBank mRNA | BC155331.1 | - | 11 - 12 | 2030 | 94 | |
More Information
Set Membership: |
|

shrprbck1r
Genomic Map