Assay Details
| Gene Symbol: shrprbck1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111200.1 | NP_001104670.1 | 3 - 4 | 743 | 75 | |
| XM_005166435.4 | XP_005166492.1 | 3 - 4 | 782 | 75 | ||
| GenBank mRNA | BC155331.1 | - | 3 - 4 | 743 | 75 | |
More Information
Set Membership: |
|

shrprbck1r
Genomic Map