Assay Details
| Gene Symbol: ca5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111201.1 | NP_001104671.1 | 1 - 2 | 549 | 119 | |
| GenBank mRNA | BC153994.1 | - | 1 - 2 | 549 | 119 | |
More Information
Set Membership: |
|

ca5a
Genomic Map