Assay Details
| Gene Symbol: ca5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111201.1 | NP_001104671.1 | 5 - 6 | 1025 | 132 | |
| XM_005163076.4 | XP_005163133.1 | 5 - 6 | 858 | 132 | ||
| GenBank mRNA | BC153994.1 | - | 5 - 6 | 1025 | 132 | |
More Information
Set Membership: |
|

ca5a
Genomic Map