Assay Details
| Gene Symbol: slc35f3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320053.1 | NP_001306982.1 | 5 - 6 | 1556 | 68 | |
| GenBank mRNA | BC154016.1 | - | 5 - 6 | 944 | 68 | |
More Information
Set Membership: |
|

slc35f3a
Genomic Map