Assay Details
| Gene Symbol: dkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111209.1 | NP_001104679.1 | 3 - 4 | 812 | 130 | |
| GenBank mRNA | AB753454.1 | - | 3 - 4 | 736 | 130 | |
| BC153973.1 | - | 3 - 4 | 812 | 130 | ||
More Information
Set Membership: |
|

dkk2
Genomic Map