Assay Details
Gene Symbol: hbegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111226.1 | NP_001104696.1 | 1 - 2 | 151 | 141 | |
GenBank mRNA | BC155826.1 | - | 1 - 2 | 151 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC155826.1, NM_001111226.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
