Assay Details
| Gene Symbol: myh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112733.1 | NP_001106204.1 | 32 - 33 | 4723 | 100 | |
| GenBank mRNA | AF114427.2 | - | 32 - 33 | 4729 | 100 | |
More Information
Set Membership: |
|

myh7
Genomic Map