Assay Details
| Gene Symbol: lnx2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113225.2 | NP_001106696.2 | 1 - 2 | 130 | 61 | |
| XM_021470171.1 | XP_021325846.1 | 1 - 2 | 133 | 61 | ||
| GenBank mRNA | EB950577.1 | - | 1 - 2 | 78 | 61 | |
| EH583415.1 | - | 1 - 2 | 141 | 61 | ||
| FJ156085.1 | - | 1 - 2 | 130 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001113225.2, XM_021470171.1 transcripts.
Set Membership: |
|

lnx2a
Genomic Map