Assay Details
| Gene Symbol: lnx2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113225.2 | NP_001106696.2 | 5 - 6 | 1205 | 127 | |
| XM_021470169.1 | XP_021325844.1 | 4 - 5 | 695 | 127 | ||
| XM_021470170.1 | XP_021325845.1 | 4 - 5 | 603 | 127 | ||
| XM_021470171.1 | XP_021325846.1 | 5 - 6 | 1208 | 127 | ||
| GenBank mRNA | EB957408.1 | - | 2 - 3 | 231 | 127 | |
| FJ156085.1 | - | 5 - 6 | 1205 | 127 | ||
More Information
Set Membership: |
|

lnx2a
Genomic Map