Assay Details
| Gene Symbol: slc9a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113476.1 | NP_001106947.1 | 9 - 10 | 1202 | 82 | |
| GenBank mRNA | EF591986.1 | - | 9 - 10 | 1202 | 82 | |
More Information
Set Membership: |
|

slc9a6b
Genomic Map