Assay Details
Gene Symbol: keap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113477.1 | NP_001106948.1 | 2 - 3 | 579 | 67 | |
XM_005168685.3 | XP_005168742.1 | 2 - 3 | 727 | 67 | ||
XM_021476739.1 | XP_021332414.1 | 2 - 3 | 617 | 67 | ||
GenBank mRNA | AB271119.1 | - | 2 - 3 | 579 | 67 | |
BC155079.1 | - | 2 - 3 | 603 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
