Assay Details
| Gene Symbol: slc9a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113480.1 | NP_001106952.1 | 6 - 7 | 1418 | 83 | |
| XM_021467239.1 | XP_021322914.1 | 6 - 7 | 1423 | 83 | ||
| XM_021467240.1 | XP_021322915.1 | 5 - 6 | 1188 | 83 | ||
| GenBank mRNA | EF591982.1 | - | 6 - 7 | 1418 | 83 | |
More Information
Set Membership: |
|

slc9a1b
Genomic Map