Assay Details
| Gene Symbol: prlrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113500.1 | NP_001106972.1 | 4 - 5 | 559 | 95 | |
| XM_005165610.3 | XP_005165667.1 | 5 - 6 | 765 | 95 | ||
| GenBank mRNA | EU517720.1 | - | 5 - 6 | 737 | 95 | |
More Information
Set Membership: |
|

prlrb
Genomic Map