Assay Details
| Gene Symbol: il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113507.2 | NP_001106979.2 | 1 - 2 | 174 | 65 | |
| GenBank mRNA | CN018734.1 | - | 1 - 2 | 197 | 65 | |
More Information
Set Membership: |
|

il7r
Genomic Map