Assay Details
| Gene Symbol: cadm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113555.1 | NP_001107027.1 | 4 - 5 | 389 | 73 | |
| XM_005157473.4 | XP_005157530.1 | 4 - 5 | 706 | 73 | ||
| XM_005157474.4 | XP_005157531.1 | 4 - 5 | 706 | 73 | ||
| XM_005157475.4 | XP_005157532.1 | 4 - 5 | 705 | 73 | ||
| XM_005157476.4 | XP_005157533.1 | 4 - 5 | 707 | 73 | ||
| XM_009291496.3 | XP_009289771.1 | 4 - 5 | 705 | 73 | ||
| GenBank mRNA | EU182353.1 | - | 4 - 5 | 389 | 73 | |
More Information
Set Membership: |
|

cadm2b
Genomic Map