Assay Details
| Gene Symbol: cadm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113555.1 | NP_001107027.1 | 5 - 6 | 524 | 58 | |
| XM_005157473.4 | XP_005157530.1 | 5 - 6 | 841 | 58 | ||
| XM_005157475.4 | XP_005157532.1 | 5 - 6 | 840 | 58 | ||
| XM_005157476.4 | XP_005157533.1 | 5 - 6 | 842 | 58 | ||
| GenBank mRNA | EU182353.1 | - | 5 - 6 | 524 | 58 | |
More Information
Set Membership: |
|

cadm2b
Genomic Map