Assay Details
| Gene Symbol: kif22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113591.1 | NP_001107063.1 | 9 - 10 | 1642 | 61 | |
| GenBank mRNA | BC154464.1 | - | 9 - 10 | 1642 | 61 | |
| BC171488.1 | - | 9 - 10 | 1542 | 61 | ||
| BC171492.1 | - | 9 - 10 | 1542 | 61 | ||
More Information
Set Membership: |
|

kif22
Genomic Map