Assay Details
| Gene Symbol: ccl20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113595.1 | NP_001107067.1 | 2 - 3 | 174 | 118 | |
| GenBank mRNA | AB331755.1 | - | 2 - 3 | 174 | 118 | |
More Information
Set Membership: |
|

ccl20b
Genomic Map