Assay Details
| Gene Symbol: paox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113603.1 | NP_001107075.1 | 4 - 5 | 875 | 78 | |
| XM_005166871.4 | XP_005166928.1 | 6 - 7 | 1244 | 78 | ||
More Information
Set Membership: |
|

paox1
Genomic Map