Assay Details
| Gene Symbol: gkup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113616.1 | NP_001107088.1 | 5 - 6 | 525 | 126 | |
| XM_005157527.4 | XP_005157584.1 | 5 - 6 | 592 | 126 | ||
| GenBank mRNA | BC154511.1 | - | 5 - 6 | 525 | 126 | |
| BC171705.1 | - | 4 - 5 | 328 | 126 | ||
| BC171707.1 | - | 4 - 5 | 328 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC154511.1, BC171705.1, BC171707.1, NM_001113616.1 transcripts.
Set Membership: |
|

gkup
Genomic Map