Assay Details
| Gene Symbol: sgk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113622.1 | NP_001107094.1 | 2 - 3 | 201 | 94 | |
| XM_009296592.3 | XP_009294867.1 | 2 - 3 | 261 | 94 | ||
| GenBank mRNA | BC154535.1 | - | 2 - 3 | 201 | 94 | |
| BC171486.1 | - | 2 - 3 | 168 | 94 | ||
| BC171490.1 | - | 2 - 3 | 168 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC154535.1, BC171486.1, BC171490.1, NM_001113622.1, XM_009296592.3 transcripts.
Set Membership: |
|

sgk2a
Genomic Map