Assay Details
| Gene Symbol: sgk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113622.1 | NP_001107094.1 | 4 - 5 | 313 | 76 | |
| XM_009296592.3 | XP_009294867.1 | 4 - 5 | 373 | 76 | ||
| GenBank mRNA | BC154535.1 | - | 4 - 5 | 313 | 76 | |
| BC171486.1 | - | 4 - 5 | 280 | 76 | ||
| BC171490.1 | - | 4 - 5 | 280 | 76 | ||
More Information
Set Membership: |
|

sgk2a
Genomic Map