Assay Details
| Gene Symbol: ccn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113628.1 | NP_001107100.1 | 1 - 2 | 183 | 71 | |
| GenBank mRNA | BC154630.1 | - | 1 - 2 | 183 | 71 | |
| BC171541.1 | - | 1 - 2 | 179 | 71 | ||
| BC171543.1 | - | 1 - 2 | 179 | 71 | ||
| GQ273496.1 | - | 1 - 2 | 242 | 71 | ||
More Information
Set Membership: |
|

ccn4b
Genomic Map