Assay Details
| Gene Symbol: map7d1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113635.1 | NP_001107107.1 | 3 - 4 | 342 | 76 | |
| XM_005173747.1 | XP_005173804.1 | 3 - 4 | 610 | 76 | ||
| GenBank mRNA | BC154607.1 | - | 3 - 4 | 342 | 76 | |
More Information
Set Membership: |
|

map7d1b
Genomic Map