Assay Details
| Gene Symbol: map7d1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113635.1 | NP_001107107.1 | 5 - 6 | 620 | 66 | |
| XM_005173748.1 | XP_005173805.1 | 6 - 7 | 904 | 66 | ||
| GenBank mRNA | BC154607.1 | - | 5 - 6 | 620 | 66 | |
More Information
Set Membership: |
|

map7d1b
Genomic Map