Assay Details
| Gene Symbol: map7d1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113635.1 | NP_001107107.1 | 6 - 7 | 738 | 98 | |
| XM_005173746.1 | XP_005173803.1 | 8 - 9 | 1127 | 98 | ||
| XM_005173747.1 | XP_005173804.1 | 7 - 8 | 1111 | 98 | ||
| XM_005173748.1 | XP_005173805.1 | 7 - 8 | 1022 | 98 | ||
| XM_009293978.3 | XP_009292253.1 | 8 - 9 | 1127 | 98 | ||
| GenBank mRNA | BC154607.1 | - | 6 - 7 | 738 | 98 | |
More Information
Set Membership: |
|

map7d1b
Genomic Map