Assay Details
| Gene Symbol: sparta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113658.1 | NP_001107130.1 | 1 - 2 | 407 | 63 | |
| XM_005172634.4 | XP_005172691.1 | 1 - 2 | 684 | 63 | ||
| GenBank mRNA | BC154514.1 | - | 1 - 2 | 407 | 63 | |
More Information
Set Membership: |
|

sparta
Genomic Map