Assay Details
| Gene Symbol: sparta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113658.1 | NP_001107130.1 | 3 - 4 | 993 | 82 | |
| XM_005172634.4 | XP_005172691.1 | 3 - 4 | 1270 | 82 | ||
| GenBank mRNA | BC154514.1 | - | 3 - 4 | 993 | 82 | |
More Information
Set Membership: |
|

sparta
Genomic Map