Assay Details
| Gene Symbol: lifrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113732.1 | NP_001107204.1 | 12 - 13 | 1818 | 79 | |
| XM_009295324.3 | XP_009293599.1 | 12 - 13 | 1888 | 79 | ||
More Information
Set Membership: |
|

lifrb
Genomic Map