Assay Details
| Gene Symbol: lifrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113732.1 | NP_001107204.1 | 9 - 10 | 1358 | 116 | |
| XM_009295324.3 | XP_009293599.1 | 9 - 10 | 1428 | 116 | ||
More Information
Set Membership: |
|

lifrb
Genomic Map