Assay Details
| Gene Symbol: socs5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113797.1 | NP_001107269.1 | 1 | 1545 | 57 | |
| XM_005156601.4 | XP_005156658.1 | 2 | 2151 | 57 | ||
| GenBank mRNA | EF195762.1 | - | 1 | 1545 | 57 | |
| FR750005.1 | - | 1 | 1545 | 57 | ||
More Information
Set Membership: |
|

socs5b
Genomic Map