Assay Details
| Gene Symbol: akirin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113800.2 | NP_001107272.1 | 1 - 2 | 444 | 82 | |
| GenBank mRNA | BC158177.1 | - | 1 - 2 | 377 | 82 | |
More Information
Set Membership: |
|

akirin1
Genomic Map