Assay Details
| Gene Symbol: ablim1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114316.1 | NP_001107788.1 | 11 - 12 | 1231 | 60 | |
| XM_009307552.3 | XP_009305827.1 | 11 - 12 | 3666 | 60 | ||
| XM_009307554.3 | XP_009305829.1 | 11 - 12 | 3639 | 60 | ||
| XM_009307559.3 | XP_009305834.1 | 11 - 12 | 3666 | 60 | ||
| XM_009307565.3 | XP_009305840.1 | 11 - 12 | 3639 | 60 | ||
| XM_017358665.2 | XP_017214154.1 | 11 - 12 | 3639 | 60 | ||
| XM_017358666.2 | XP_017214155.1 | 11 - 12 | 3639 | 60 | ||
| XM_017358667.2 | XP_017214156.1 | 11 - 12 | 3638 | 60 | ||
| XM_021480476.1 | XP_021336151.1 | 12 - 13 | 3712 | 60 | ||
| XM_021480477.1 | XP_021336152.1 | 12 - 13 | 3712 | 60 | ||
| XM_021480478.1 | XP_021336153.1 | 12 - 13 | 3684 | 60 | ||
| XM_021480479.1 | XP_021336154.1 | 12 - 13 | 3711 | 60 | ||
| XM_021480480.1 | XP_021336155.1 | 12 - 13 | 3711 | 60 | ||
| XM_021480481.1 | XP_021336156.1 | 12 - 13 | 3711 | 60 | ||
| XM_021480482.1 | XP_021336157.1 | 12 - 13 | 3711 | 60 | ||
| XM_021480483.1 | XP_021336158.1 | 12 - 13 | 3711 | 60 | ||
| GenBank mRNA | BC155652.1 | - | 11 - 12 | 1231 | 60 | |
More Information
Set Membership: |
|

ablim1a
Genomic Map