Assay Details
| Gene Symbol: ablim1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114316.1 | NP_001107788.1 | 15 - 16 | 1512 | 90 | |
| XM_021480481.1 | XP_021336156.1 | 16 - 17 | 3992 | 90 | ||
| GenBank mRNA | BC155652.1 | - | 15 - 16 | 1512 | 90 | |
More Information
Set Membership: |
|

ablim1a
Genomic Map