Assay Details
| Gene Symbol: ablim1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114316.1 | NP_001107788.1 | 18 - 19 | 1769 | 79 | |
| XM_009307552.3 | XP_009305827.1 | 19 - 20 | 4357 | 79 | ||
| XM_009307554.3 | XP_009305829.1 | 19 - 20 | 4330 | 79 | ||
| XM_009307559.3 | XP_009305834.1 | 18 - 19 | 4210 | 79 | ||
| XM_009307565.3 | XP_009305840.1 | 19 - 20 | 4324 | 79 | ||
| XM_017358665.2 | XP_017214154.1 | 18 - 19 | 4246 | 79 | ||
| XM_017358666.2 | XP_017214155.1 | 18 - 19 | 4240 | 79 | ||
| XM_017358667.2 | XP_017214156.1 | 17 - 18 | 4098 | 79 | ||
| XM_021480476.1 | XP_021336151.1 | 20 - 21 | 4403 | 79 | ||
| XM_021480477.1 | XP_021336152.1 | 20 - 21 | 4385 | 79 | ||
| XM_021480478.1 | XP_021336153.1 | 20 - 21 | 4375 | 79 | ||
| XM_021480479.1 | XP_021336154.1 | 19 - 20 | 4300 | 79 | ||
| XM_021480480.1 | XP_021336155.1 | 19 - 20 | 4255 | 79 | ||
| XM_021480481.1 | XP_021336156.1 | 19 - 20 | 4249 | 79 | ||
| XM_021480482.1 | XP_021336157.1 | 18 - 19 | 4171 | 79 | ||
| XM_021480483.1 | XP_021336158.1 | 18 - 19 | 4165 | 79 | ||
| GenBank mRNA | BC155652.1 | - | 18 - 19 | 1769 | 79 | |
More Information
Set Membership: |
|

ablim1a
Genomic Map