Assay Details
| Gene Symbol: ablim1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114316.1 | NP_001107788.1 | 22 - 23 | 1992 | 74 | |
| XM_009307552.3 | XP_009305827.1 | 23 - 24 | 4580 | 74 | ||
| XM_009307554.3 | XP_009305829.1 | 23 - 24 | 4553 | 74 | ||
| XM_009307559.3 | XP_009305834.1 | 22 - 23 | 4433 | 74 | ||
| XM_009307565.3 | XP_009305840.1 | 23 - 24 | 4547 | 74 | ||
| XM_017358665.2 | XP_017214154.1 | 22 - 23 | 4469 | 74 | ||
| XM_017358666.2 | XP_017214155.1 | 22 - 23 | 4463 | 74 | ||
| XM_017358667.2 | XP_017214156.1 | 21 - 22 | 4321 | 74 | ||
| XM_021480476.1 | XP_021336151.1 | 24 - 25 | 4626 | 74 | ||
| XM_021480477.1 | XP_021336152.1 | 24 - 25 | 4608 | 74 | ||
| XM_021480478.1 | XP_021336153.1 | 24 - 25 | 4598 | 74 | ||
| XM_021480479.1 | XP_021336154.1 | 23 - 24 | 4523 | 74 | ||
| XM_021480480.1 | XP_021336155.1 | 23 - 24 | 4478 | 74 | ||
| XM_021480481.1 | XP_021336156.1 | 23 - 24 | 4472 | 74 | ||
| XM_021480482.1 | XP_021336157.1 | 22 - 23 | 4394 | 74 | ||
| XM_021480483.1 | XP_021336158.1 | 22 - 23 | 4388 | 74 | ||
| GenBank mRNA | BC155652.1 | - | 22 - 23 | 1992 | 74 | |
More Information
Set Membership: |
|

ablim1a
Genomic Map